Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BST2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$206.50 - $517.59
Specifications
| Antigen | BST2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 - 4 ug/mL, Simple Western 1:200, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/mL, Immunohistochemistry-Frozen reported by customer review, Flow (Cell Surface) reported in scientific literature (PMID 21695198), Immunofluorescence |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227154SS
![]() |
Novus Biologicals
NBP227154SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227154
![]() |
Novus Biologicals
NBP227154 |
0.1 mg |
Each for $517.59
|
|
|||||
Description
BST2 Polyclonal specifically detects BST2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Immunofluorescence.Specifications
| BST2 | |
| Western Blot 2 - 4 ug/mL, Simple Western 1:200, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/mL, Immunohistochemistry-Frozen reported by customer review, Flow (Cell Surface) reported in scientific literature (PMID 21695198), Immunofluorescence | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| Q10589 | |
| 684 | |
| A portion of amino acids 80-130 of human BST2 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN | |
| BST2 | |
| IgG | |
| Protein A purified | |
| The BST2 amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, orangutan, white-cheeked gibbon, great ape, dwarf chimpanzee, gibbon, and 94% homologous in rhesus monkey. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title