Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTAF1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291790 Supplier Novus Biologicals Supplier No. NB10057504PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTAF1 Polyclonal antibody specifically detects BTAF1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen BTAF1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias ATP-dependent helicase BTAF1, BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1homolog, S. cerevisiae), B-TFIID transcription factor-associated 170 kDa subunit, EC 3.6.1, EC 3.6.4.-, MOT1MGC138406, TAF(II)170KIAA0940, TAF-172, TAF172BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170 kD (Mot1homolog, S. cerevisiae), TAFII170TATA-binding protein-associated factor 172, TBP-associated factor 172
Host Species Rabbit
Immunogen Maps to a region between residue 1800 and the C-terminus (residue 1849) of human RNA polymerase II, B-TFIID transcription factor-associated, 170kDa using the numbering given in entry NP_003963.1 (GeneID 9044).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9044
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.