Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD12 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP128680A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP128680A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP128680A32 Supplier Novus Biologicals Supplier No. NBP128680AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTBD12 Polyclonal specifically detects BTBD12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen BTBD12
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BTB (POZ) domain containing 12, BTB/POZ domain-containing protein 12, BTBD12structure-specific endonuclease subunit SLX4, KIAA1784MUS312, KIAA1987FANCP, SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae)
Gene Symbols SLX4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1784 to 1834 of human BTB (POZ) domain containing 12 using the numbering given in entry NP_115820.2 (GeneID 84464).
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84464.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.