Learn More
Description
Specifications
Specifications
| Antigen | BTBD12 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot reported in scientific literature (PMID 24012755), Chromatin Immunoprecipitation reported in scientific literature (PMID 24012755), Immunohistochemistry 1:250 - 1:500, Immunocytochemistry/Immunofluorescence 5 - 10 ug/ml. Use reported in scientific literature (PMID 24012755), Immunoprecipitation 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:250 - 1:500, Chromatin Immunoprecipitation (ChIP) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q8IY92 |
| Gene Alias | BTB (POZ) domain containing 12, BTB/POZ domain-containing protein 12, BTBD12structure-specific endonuclease subunit SLX4, KIAA1784MUS312, KIAA1987FANCP, SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae) |
| Gene Symbols | SLX4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
