Learn More
Description
Specifications
Specifications
| Antigen | BTBD12 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | BTB (POZ) domain containing 12, BTB/POZ domain-containing protein 12, BTBD12structure-specific endonuclease subunit SLX4, KIAA1784MUS312, KIAA1987FANCP, SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae) |
| Gene Symbols | SLX4 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 1784 to 1834 of human BTB (POZ) domain containing 12 using the numbering given in entry NP_115820.2 (GeneID 84464). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
