Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTF Antibody (M33-P5B11), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP250118IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250118IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250118IR Supplier Novus Biologicals Supplier No. NBP250118IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTF Monoclonal specifically detects BTF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen BTF
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M33-P5B11
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1
Gene Symbols BCLAF1
Host Species Mouse
Immunogen Peptide Sequence - KYQGDGIVED (amino acids 900 - 909)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9774
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.