Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTG3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB332061 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332061 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332061 Supplier Novus Biologicals Supplier No. NBP297381APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTG3 Polyclonal antibody specifically detects BTG3 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen BTG3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Abundant in neuroepithelium area protein, ANATOB55, B-cell translocation gene 3, B-cell translocation gene10, BTG family member 3, BTG family, member 3, MGC8928, protein BTG3, Protein Tob5, tob55, TOB5TOFA
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human BTG3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10950
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.