Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158173
Description
BTG4 Polyclonal specifically detects BTG4 in Human samples. It is validated for Western Blot.Specifications
| BTG4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| B-cell translocation gene 4, BTG family member 4, MGC33003, PC3BProtein PC3b, protein BTG4 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Bovine: 92%; Chicken: 85%; Mouse: 85%; Rat: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NY30 | |
| BTG4 | |
| Synthetic peptides corresponding to BTG4(B-cell translocation gene 4) The peptide sequence was selected from the middle region of BTG4. Peptide sequence ILERACVESNVDFSHLGLPKEMTIWVDPFEVCCRYGEKNHPFTVASFKGR. | |
| 100 μL | |
| Cell Cycle and Replication | |
| 54766 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction