Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTK Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357832 Supplier Novus Biologicals Supplier No. NBP176596MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTK Polyclonal antibody specifically detects BTK in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen BTK
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 16 amino acids near the amino-terminus of human BTK. The immunogen is located within amino acids 20 - 70 of BTK. Amino Acid Squence: SYYEYDFERGRRGSKK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 695
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.