Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA/CD272 Antibody (922701), Biotin, Novus Biologicals™
SDP

Catalog No. MAB33541B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB33541B 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB33541B Supplier Novus Biologicals Supplier No. MAB33541B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BTLA/CD272
Applications Flow Cytometry
Classification Monoclonal
Clone 922701
Conjugate Biotin
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Gene Symbols BTLA
Host Species Mouse
Immunogen rhBTLA aa25-150
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151888
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.