Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA/CD272 Antibody (BTLA/1528), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB379477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379477 Supplier Novus Biologicals Supplier No. NBP308619C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human samples. It is validated for ELISA

Specifications

Antigen BTLA/CD272
Applications ELISA
Classification Monoclonal
Clone BTLA/1528
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Host Species Mouse
Immunogen Recombinant extracellular human BTLA/CD272 protein (Uniprot: Q7Z69)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 151888
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.