Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA/CD272 Antibody (HMBT-6B2), HRP, Novus Biologicals™
SDP

Catalog No. NB016491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016491 Supplier Novus Biologicals Supplier No. NBP281117H
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Mouse samples. It is validated for Western Blot, Immunoblotting.

Specifications

Antigen BTLA/CD272
Applications Western Blot, Immunoblot
Classification Monoclonal
Clone HMBT-6B2
Conjugate HRP
Dilution Western Blot, Immunoblotting
Formulation PBS
Gene Alias B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Gene Symbols BTLA
Host Species Armenian Hamster
Immunogen This antibody was raised by immunising Armenian hamsters with mouse BTLA/CD272:Fc fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 151888
Test Specificity This antibody is specific for B and T lymphocyte attenuator (BTLA), which functions as a negative regulator of T cell activation and proliferation, and attenuates B cell proliferation upon associating with its known ligand, herpes virus entry mediator (HVEM).
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.