Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA/CD272 Antibody (MIH26), PerCP, Novus Biologicals™
SDP

Catalog No. NB158443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158443 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB158443 Supplier Novus Biologicals Supplier No. NBP314617PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human, Non-human Primate samples. It is validated for Flow Cytometry

Specifications

Antigen BTLA/CD272
Applications Flow Cytometry
Classification Monoclonal
Clone MIH26
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Host Species Mouse
Immunogen Cells transfected with human BTLA/CD272
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 151888
Target Species Human, Non-human Primate
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.