Learn More
BTLA/CD272, Armenian Hamster anti-Mouse, Clone: HMBT-6B2, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | BTLA/CD272 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, Immunoblot, Blocking Assay |
| Classification | Monoclonal |
| Clone | HMBT-6B2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, Immunoprecipitation, Functional, Immunoblotting, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 |
| Gene Symbols | BTLA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.