Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTN1A1/Butyrophilin Antibody (2151C), DyLight 550, R&D Systems™
SDP

Catalog No. MAB8467R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8467R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8467R Supplier Novus Biologicals Supplier No. MAB8467R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BTN1A1/Butyrophilin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2151C
Conjugate DyLight 550
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1
Gene Symbols BTN1A1
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 696
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.