Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTN3A1/2/3 Antibody (849203) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB375120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375120 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375120 Supplier Novus Biologicals Supplier No. FAB7136J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTN3A1/2/3 Monoclonal antibody specifically detects BTN3A1/2/3 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen BTN3A1/2/3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 849203
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BT3.1;BTF5;BTN3.1;BTN3A1;CD277
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD277/BTN3A1, Met1-Gly254, Accession # O00481
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.