Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTN3A3 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB091149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB091149 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB091149 Supplier Novus Biologicals Supplier No. NBP300127AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTN3A3 Polyclonal specifically detects BTN3A3 in Human samples. It is validated for ELISA.

Specifications

Antigen BTN3A3
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias BTF3butyrophilin 3, butyrophilin subfamily 3 member A3, butyrophilin, subfamily 3, member A3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant BTN3A3 (O00478-1; Met1-Trp248).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10384
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.