Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTNL8 Antibody (2187B), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345757 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345757 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345757 Supplier Novus Biologicals Supplier No. FAB9359MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BTNL8 Monoclonal specifically detects BTNL8 in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen BTNL8
Applications Flow Cytometry, Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2187B
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK-293-derived Human BTNL8, Gln18-Lys238, Accession # Q6UX41-1
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79908
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.