Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bub3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB357381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357381 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357381 Supplier Novus Biologicals Supplier No. NBP176796MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bub3 Polyclonal antibody specifically detects Bub3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Bub3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human bub3. The immunogen is located within the last 50 amino acids of Bub3. Amino Acid Squence: SYMYEMDDTEHPEDG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9184
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.