Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUD31 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB326955 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326955 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326955 Supplier Novus Biologicals Supplier No. NBP299132F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BUD31 Polyclonal antibody specifically detects BUD31 in Human samples. It is validated for Western Blot

Specifications

Antigen BUD31
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias BUD31 homolog (S. cerevisiae), BUD31 homolog (yeast), EDG-2, EDG2MGC111202, fSAP17, functional spliceosome-associated protein 17, G10, G10 maternal transcript homolog, maternal G10 transcript, protein BUD31 homolog, Protein EDG-2, Protein G10 homolog, YCR063W
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human BUD31.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 8896
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.