Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
10 L
6 x 1 L
Description
Leading the Industry For Over 65 Years. Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured.
Composition per liter:
- 3.52 g Potassium dihydrogen phosphate (KH2PO4)
- 7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
Specifications
Specifications
| pH | 7.0 |
| Quantity | 500 mL |
| MDL Number | MFCD00066650 |
| UN Number | NONH for all modes of transport |
| IUPAC Name | azanium;hydroxide |
| Chemical Name or Material | Buffer solution |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction