Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphate Buffer, pH 7, for Residual Chlorine and Chlorine Dioxide (Amperometric), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, Phosphate |
| CAS | 7732-18-5, 7558-79-4, 7778-77-0 |
| For Use With (Application) | APHA for Chlorine and Chlorine Dioxide (Amperometric Method) |
| pH | 7 |
| Physical Form | Solution |
| Quantity | 1 L |
| Packaging | Natural Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction