Learn More
Bullard™ Inlet and Outlet Filters for Electric Free-Air™ Pumps
SDP
Catalog No. 19021342 Shop All Bullard Company Products
Click to view available options
Product Type:
Inlet filter
Outlet filter
For use with Free-Air pumps
Coupler Splitter/Adapter: Converts coupler to double 1/2 in. Industrial Interchange female couplers (one flow through, one shut-off)
Inlet Filter: Change every 500 hours or as required
Outlet Filter: Carbofine; change every 200 hours or as required
Specifications
Model 23611
Product Type Inlet filter
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.