Learn More
Bullard™ Free-Air EDP30 Electric Driven Pumps
SDP
Catalog No. 17604205 Shop All Bullard Company Products
Supports climate control devices
Shipped complete with inlet and outlet filter cartridges, pressure gauges, pressure relief valves, and one air supply hose quick-disconnect pod with six 0.5 in. Industrial Interchange (Hansen compatible) quick-disconnect couplers.
  • Supplies air for four hood-style, six half or full face mask airline respirators, or two hood-style respirators equipped with Frigitron climate control devices
  • Respirators must be approved to operate on 30 psig or less
  • Easy operation and low maintenance
  • Completely mobile and portable
  • No expensive installation costs
Specifications
Applications Support Climate Control Devices
Includes Inlet and outlet filter cartridge, pressure gauges, pressure relief valves and one air supply hose quick-disconnect pod with six 0.5 in. industrial interchange quick-disconnect couplers, reversing starter
Product Type Electric Driven Air Pump
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.