Learn More
Bullard™ Vector Cap-Style Helmet
SDP
Complies with ANSI/ISEA Z89.1-2009 Type II, as well as CSA Z94.1-2005, Type II head protection standards
$72.80
Products 5
Catalog Number Mfr. No. Color Suspension Type Price Quantity & Availability  
Catalog Number Mfr. No. Color Suspension Type Price Quantity & Availability  
19-098-036
SDP
View Documents
Bullard™
VTHGR
Hi-vis Green Ratchet
Each for $72.80
Case of 10 EA for $355.00
Only null left
Add to Cart
19-098-038
SDP
View Documents
Bullard™
VTHYR
Hi-vis Yellow Ratchet
Each for $72.80
Only null left
Add to Cart
19-098-035
SDP
View Documents
Bullard™
VTHYP
Hi-vis Yellow Pin-Lock N/A N/A N/A
19-098-033
SDP
missing translation for 'viewDocuments'
Bullard™
VTHGP
Hi-vis Green Pin-Lock N/A N/A N/A
19-098-034
SDP
missing translation for 'viewDocuments'
Bullard™
VTORP
Hi-vis Orange Pin-Lock N/A N/A N/A
Description
Description
Provides impact protection from all directions: top, front, sides and rear
  • 4-point suspension
  • Replaceable, moisture wicking Sportek™ brow pad
  • Built-in goggle strap retaining slots
  • Polyethylene outer shell
  • EPS foam impact liner
Order Info
Available in either 4-point Flex-Gear™ or self-sizing suspension
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.