Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Loose Flame Resistant Jean, Stonewash
Shop All Red Kap Industries Products
Click to view available options
Size:
28 x 30 in.
28 x 32 in.
28 x 34 in.
30 x 30 in.
30 x 32 in.
30 x 34 in.
31 x 30 in.
31 x 32 in.
31 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Pre-Washed Cotton Denim
- Closure: One-piece waistband with metal button closure (metal is not exposed to the skin)
- Pocket: Five jean-style pockets
- Features:
- Sits below natural waist
- Loose seat and thigh with wider leg opening than our classic fit jean
- Soft pre-wash and stone-washed fabric for comfort and look
- Product Collection: Excel FR, Denim
- Packaging: Waistband ticket
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits just below the waist. Extra room in the seat and thigh. Fuller in the leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
Size | 33 x 32 in. |
Gender | Men's |
Product Type | Flame-resistant Jeans |
Material | Denim |
Color | Blue |
No. of Pockets | 5 |
Closure Type | Zipper |
Resistant To | Flame |
Hazard Risk Category | HRC2 |
Arc Flash Rating | ATPV 20.7 Calories/cm2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction