Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VF Workwear Bulwark Women's Flame Resistant Curvy Fit Denim Jeans
Shop All Red Kap Industries Products
Click to view available options
Size (English) Inseam:
28 in.
30 in.
32 in.
34 in.
Size:
6
8
10
12
14
Specifications
Specifications
Size | 10 |
Gender | Women's |
Material | Cotton, Modacrylic |
Color | Blue |
No. of Pockets | 5 |
Closure Type | Button |
Hazard Risk Category | HRC2 |
Arc Flash Rating | EBT 13.0 Calories/cm2 |
Size (English) Inseam | 30 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction