Learn More
United Scientific™ Burette Clamp
Shop All United Scientific Supplies, Inc Products
Click to view available options
Type:
Burette Clamp (Double)
Burette Clamp (Single)
Description
- These polypropylene burette clamps are completely corrosion-proof.
- The spring-loaded jack in the middle provides a vice like grip on the burette and prevents it from slipping.
- Two tapered wedges are included to ensure firm grip on 1/2 or 3/8 in. diameter rods.
Specifications
Specifications
Type | Burette Clamp (Single) |
Material | Polypropylene |
Color | White |
Jaw Type | Spring Loaded |