Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Busulfan, 98%

Catalog No. aaj6134806
Click to view available options
Quantity:
5 g

A cell cycle non-specific alkylating antineoplastic agent | CAS: 55-98-1 | C6H14O6S2 | 246.292 g/mol

A cell cycle non-specific alkylating antineoplastic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55-98-1
Color Brown
Molecular Formula C6H14O6S2
Linear Formula CH3SO2O(CH2)4OSO2CH3
Quantity 5 g
MDL Number MFCD00007562
UN Number UN2811
Beilstein 1791786
Merck Index 14,1505
Synonym busulfan, myleran, busulphan, sulphabutin, leucosulfan, myelosan, busulfex, citosulfan, mielucin, misulban
InChI Key COVZYZSDYWQREU-UHFFFAOYSA-N
SMILES CS(=O)(=O)OCCCCOS(=O)(=O)C
IUPAC Name 4-methylsulfonyloxybutyl methanesulfonate
Molecular Weight (g/mol) 246.292
PubChem CID 2478
ChEBI CHEBI:28901
Formula Weight 246.29
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Busulfan
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335-H340-H350-H361d
Hazard Statement GHS H Statement
H300-H310-H330-H350
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 200-250-2
RTECSNumber EK1750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.