Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyraldehyde Dibutyl Acetal 98.0+%, TCI America™
SDP

Catalog No. B421825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B421825G 25 g
B42185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B421825G Supplier TCI America Supplier No. B421825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5921-80-2
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.338
MDL Number MFCD00561028
InChI Key WQRBJFZKPWPIJD-UHFFFAOYSA-N
Synonym 1,1-Dibutoxybutane
PubChem CID 22210
IUPAC Name 1,1-dibutoxybutane
SMILES CCCCOC(CCC)OCCCC

Specifications

Color Colorless
Boiling Point 214°C
Quantity 25 g
UN Number 1993
Formula Weight 202.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyraldehyde Dibutyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.