Alfa Aesar™ Butyric anhydride, 98%

Manufacturer: Alfa Aesar™ A12377AE

Quantity 100mL

 View more versions of this product

Catalog No. AAA12377AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106-31-0
    Molecular Formula C8H14O3
    Linear Formula (CH3CH2CH2CO)2O
    Formula Weight 158.2
    MDL Number MFCD00009389
    UN Number UN2739
    Beilstein 1099474
    Merck Index 14,1594
    Chemical Name or Material Butyric anhydride

    Specifications

    Formula Weight 158.2
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Molecular Formula C8H14O3
    CAS 106-31-0
    Merck Index 14,1594
    Melting Point −75°C
    Health Hazard 1 Danger
    RTECS Number ET7090000
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Flash Point 88°C (190°F)
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYRIC ANHYDRIDE
    Linear Formula (CH3CH2CH2CO)2O
    MDL Number MFCD00009389
    UN Number UN2739
    Beilstein 1099474
    Boiling Point 198° to 199°C
    Quantity 100mL
    Odor Rancid
    Chemical Name or Material Butyric anhydride
    Assay 98%

    Description

    Applications
    Butyric anhydride is used in the preparation of amidoamine dendron-based co-adsorbents, which finds application in dye- sensitized solar cells improvement. It is also used in the synthesis of butyrate ester, which is used as a perfume and flavor. Further, it acts as a fumigant to drive bees from their hives. In addition to this, it is used in food additives, textile auxiliaries, varnishes, perfumes, pharmaceuticals and disinfectants.

    Solubility
    Miscible with water and ether.

    Notes
    Air sensitive. Incompatible with strong oxidizing agents.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit