Learn More
Butyryl chloride, 98%
Description
Butyryl chloride is used as an intermediate for the synthesis of active pharmaceutical ingredients benzoyl peroxide, t-butyl perbenzoate, and benzophenone. It is also used as an acylation reagent. Further, it is used in the preparation of peroxides such as benzoyl peroxide, benzophenone and t-butyl perbenzoate. In addition to this, it is involved in the preparation of chromones and antiproliferative agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Butyryl chloride is used as an intermediate for the synthesis of active pharmaceutical ingredients benzoyl peroxide, t-butyl perbenzoate, and benzophenone. It is also used as an acylation reagent. Further, it is used in the preparation of peroxides such as benzoyl peroxide, benzophenone and t-butyl perbenzoate. In addition to this, it is involved in the preparation of chromones and antiproliferative agents.
Solubility
Miscible with ether.
Notes
Moisture sensitive. Incompatible with water, alcohols, strong bases and strong oxidizing agents
Specifications
Specifications
| Melting Point | -89°C |
| Density | 1.026 |
| Boiling Point | 101°C to 102°C |
| Flash Point | 18°C (64°F) |
| Odor | Acrid |
| Refractive Index | 1.412 |
| Quantity | 250 g |
| UN Number | UN2353 |
| Beilstein | 605395 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H314-H331 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYRYL CHLORIDE |
| EINECSNumber | 205-498-5 |
| RTECSNumber | EM0595000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.