Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyrylcholinesterase/BCHE Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB053168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053168 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053168 Supplier Novus Biologicals Supplier No. NBP298275AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Butyrylcholinesterase/BCHE Polyclonal specifically detects Butyrylcholinesterase/BCHE in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Butyrylcholinesterase/BCHE
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Acylcholine acylhydrolase, Butyrylcholine esterase, butyrylcholinesterase, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Butyrylcholinesterase/BCHE (NP_033868.3; Met1-Leu603).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 590
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.