Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyrylcholinesterase/BCHE Antibody (OTI1E1), Novus Biologicals™
SDP

Catalog No. NBP203523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP203523 Supplier Novus Biologicals Supplier No. NBP203523
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Butyrylcholinesterase/BCHE Monoclonal specifically detects Butyrylcholinesterase/BCHE in Human, Canine samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Butyrylcholinesterase/BCHE
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1E1
Concentration 0.72 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Flow Cytometry 1:100
Gene Alias Acylcholine acylhydrolase, Butyrylcholine esterase, butyrylcholinesterase, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase
Gene Symbols BCHE
Host Species Mouse
Immunogen Full length human recombinant protein of human BCHE (NP_000046) produced in HEK293T cell.
Molecular Weight of Antigen 68.2 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 590
Target Species Human, Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.