Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUV805 Rat Anti-Mouse CD274 (PD-L1), BD Horizon™
Rat Monoclonal Antibody
$220.00 - $448.00
Specifications
Antigen | CD274 (PD-L1) |
---|---|
Clone | MIH5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD274 (PD-L1) Monoclonal antibody specifically detects CD274 (PD-L1) in Mouse samples. It is validated for Flow cytometrySpecifications
CD274 (PD-L1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
DBA/2 mouse lymphoma L5178Y transfected with Pdcd1lg1 cDNA | |
Affinity Chromatography |
MIH5 | |
Flow Cytometry | |
Brilliant Ultraviolet 805 | |
Rat | |
Mouse | |
B7-H1, PD-L1; PD1L1; Programmed death ligand 1 | |
IgG2a λ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title