Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human Chondroitin Sulfate (NG2), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570318
Description
Chondroitin Sulfate (NG2) Monoclonal antibody specifically detects Chondroitin Sulfate (NG2) in Human samples. It is validated for Flow cytometry
Specifications
Chondroitin Sulfate (NG2) | |
Monoclonal | |
Brilliant Violet 421 | |
CSPG4; HMW-MAA; MCSP; MCSPG; NG2 | |
Human BM stromal cell line established by infection with SV-40 | |
100 Tests | |
1464 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
7.1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction