Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human Chondroitin Sulfate (NG2), BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | Chondroitin Sulfate (NG2) |
---|---|
Clone | 7.1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
Chondroitin Sulfate (NG2) Monoclonal antibody specifically detects Chondroitin Sulfate (NG2) in Human samples. It is validated for Flow cytometrySpecifications
Chondroitin Sulfate (NG2) | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
CSPG4; HMW-MAA; MCSP; MCSPG; NG2 | |
Human BM stromal cell line established by infection with SV-40 | |
Affinity Chromatography |
7.1 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
1464 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title