Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human PD-1 (CD279), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569466
Description
PD-1 (CD279) Monoclonal antibody specifically detects PD-1 (CD279) in Human samples. It is validated for Flow cytometry
Specifications
| PD-1 (CD279) | |
| Monoclonal | |
| Brilliant Violet 421 | |
| PD1; PDC1; PDCD1; Programmed cell death 1; SLEB2; hPD-1 | |
| Not Reported | |
| 25 Tests | |
| 5133 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry | |
| NAT105 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction