Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human PD-1 (CD279), BD Horizon™
Mouse Monoclonal Antibody
$232.00 - $448.00
Specifications
| Antigen | PD-1 (CD279) |
|---|---|
| Clone | NAT105 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
Description
PD-1 (CD279) Monoclonal antibody specifically detects PD-1 (CD279) in Human samples. It is validated for Flow cytometrySpecifications
| PD-1 (CD279) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Human | |
| PD1; PDC1; PDCD1; Programmed cell death 1; SLEB2; hPD-1 | |
| Not Reported | |
| Affinity Chromatography |
| NAT105 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 5133 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title