Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Mouse Anti-Human Pre-BCR, BD Horizon™

Catalog No. BDB570054 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570054 0.1 mg
BDB570135 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570054 Supplier BD Biosciences Supplier No. 570054
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pre-BCR Monoclonal antibody specifically detects Pre-BCR in Human samples. It is validated for Flow cytometry|Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pre-BCR
Applications Flow Cytometry
Classification Monoclonal
Clone HSL2
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Pre-B Cell Receptor
Host Species Mouse
Immunogen Human Pre-BCR Complexes Purified from Ltk-Pre-BCR Transfectants
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 613
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.