Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human PSMA, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570315
Description
PSMA Monoclonal antibody specifically detects PSMA in Human samples. It is validated for Flow cytometry
Specifications
PSMA | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
107-1A4 | |
Brilliant Violet 421 | |
FGCP; FOLH1; GCP2; GCPII; NAALADase I; PSM; mGCP | |
Normal Human Prostate Homogenate and LNCaP Cell Line | |
0.1 mg | |
2346 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction