Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human PSMA, BD Horizon™
Mouse Monoclonal Antibody
$214.00 - $535.00
Specifications
| Antigen | PSMA |
|---|---|
| Clone | 107-1A4 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
PSMA Monoclonal antibody specifically detects PSMA in Human samples. It is validated for Flow cytometrySpecifications
| PSMA | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 2346 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 107-1A4 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Human | |
| FGCP; FOLH1; GCP2; GCPII; NAALADase I; PSM; mGCP | |
| Normal Human Prostate Homogenate and LNCaP Cell Line | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title