Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Rat Anti-Human CAT1 (SLC7A1), BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 570128
Description
CAT1 (SLC7A1) Monoclonal antibody specifically detects CAT1 (SLC7A1) in Human samples. It is validated for Flow cytometry
Specifications
CAT1 (SLC7A1) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
CA2 | |
Brilliant Violet 421 | |
ATRC1; CAT-1; ERR; REC1L; SLC7A1; ecotropic retroviral leukemia receptor homolog | |
Human CAT1 Transfected Cells | |
25 μg | |
6541 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction