Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Rat Anti-Human CAT1 (SLC7A1), BD Horizon™
Rat Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | CAT1 (SLC7A1) |
---|---|
Clone | CA2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CAT1 (SLC7A1) Monoclonal antibody specifically detects CAT1 (SLC7A1) in Human samples. It is validated for Flow cytometrySpecifications
CAT1 (SLC7A1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
6541 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
CA2 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Rat | |
Human | |
ATRC1; CAT-1; ERR; REC1L; SLC7A1; ecotropic retroviral leukemia receptor homolog | |
Human CAT1 Transfected Cells | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title