Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Rat Anti-Mouse CD140b (PDGFRβ), BD Horizon™
Rat Monoclonal Antibody
$130.00 - $326.00
Specifications
Antigen | CD140b (PDGFRβ) |
---|---|
Clone | APB5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD140b (PDGFRβ) Monoclonal antibody specifically detects CD140b (PDGFRβ) in Mouse samples. It is validated for Flow cytometrySpecifications
CD140b (PDGFRβ) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
18596 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
APB5 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Rat | |
Mouse | |
CD140b; PDGF-R-beta; PDGFR-beta; Pdgfrb; platelet-derived growth factor receptor beta | |
Recombinant Mouse PDGFRβ Extracellular Domain Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title