Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV510 Rat Anti-Mouse CD39, BD Horizon™
Rat Monoclonal Antibody
$130.00 - $326.00
Specifications
Antigen | CD39 |
---|---|
Clone | Y23-1185 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD39 Monoclonal antibody specifically detects CD39 in Mouse samples. It is validated for Flow cytometrySpecifications
CD39 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
12495 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Y23-1185 | |
Flow Cytometry | |
Brilliant Violet 510 | |
Rat | |
Mouse | |
ectonucleoside triphosphate diphosphohydrolase 1; Entpd1; ecto-ATP diphosphohydrolase 1; ecto-ATPDase 1; ecto-ATPase 1; Cd39; NTPDase 1 | |
Mouse CD39 Transfected Cell Line | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title