Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BW-B 70C, Tocris Bioscience™

Catalog No. 130450
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Lipoxygenase inhibitor

Potent, selective inhibitor of 5-lipoxygenase. Long half-life and high oral bioavailability in vivo.

Chemical Identifiers

CAS 134470-38-5
Molecular Formula C17H17FN2O3
Molecular Weight (g/mol) 316.332
InChI Key UAIYNMRLUHHRMF-AATRIKPKSA-N
PubChem CID 5353454
ChEBI CHEBI:75307
IUPAC Name 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
SMILES CC(C=CC1=CC(=CC=C1)OC2=CC=C(C=C2)F)N(C(=O)N)O

Specifications

Quantity 50 mg
Synonym bw b70c, bwb70c, bw-b70c, bw-b 70c, chembl86676, e-n-3-3-4-fluorophenoxy phenyl-1-r,s-methylprop-2-enyl-n-hydroxyurea, n-3-3-4-fluorophenoxy phenyl-1-methyl-2-propenyl-n-hydroxyurea, urea, n-3-3-4-fluorophenoxy phenyl-1-methyl-2-propenyl-n-hydroxy, 1-3e-4-3-4-fluorophenoxy phenyl but-3-en-2-yl-1-hydroxyurea
Formula Weight 316.33
Percent Purity >98%
Chemical Name or Material BW-B 70C
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.