Learn More
Honeywell BW™ Solo, Non-Wireless Single-Gas Detector
Honeywell BW™ Solo, Non-Wireless Single-Gas Detector features BLE connectivity for real-time visibility into the status and safety of hazardous-area workers, allowing companies respond faster, more knowingly and decisively to safety incidents.
Specifications
Certifications/Compliance | Class I, Division 1, Groups A, B, C, D T4; Class I, Zone 0, AEx/Ex ia IIC T4 Ga -40°C ≤ Tamb ≤60°C; ATEX: Sira 19ATEX2004; X 0518 g II 1G Ex ia IIC T4 Ga -40°C ≤ Tamb ≤60°C;; g I M1 Ex ia I Ma -40°C ≤ Tamb ≤60°C;; IECEx: IECEx SIR 19.0004; Ex ia IIC T4 Ga / Ex ia I Ma -40°C ≤ Tamb ≤60°C;; FCC ID: SU3BWS1; IC: 20969-BWS1; RED: RE-D Directive 2014/53/EU |
---|---|
Height (Metric) | 7 cm |
Includes | Detector with specified sensor, stainless-steel alligator clip and concussion-proof housing; Test Cap with 1 ft of hose; Quick Reference Guide |
Length (Metric) | 6.7 cm |
Weight (English) | 3.6 to 4.1 oz. |
Catalog Number | Mfr. No. | Components Measured | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components Measured | Price | Quantity & Availability | |||||
19-807-573
|
Honeywell
BWSAY |
Ammonia | N/A | N/A | N/A | ||||
19-807-574
|
Honeywell
BWSRY |
Hydrogen | N/A | N/A | N/A | ||||
Description
- User-friendly operation
- Easy-to-read display for multiple languages
- Lightweight convenience
- Ease of maintenance
- Serviceability
- The new 1-Series sensor for enhanced reliability
- Connected Bluetooth Low Energy (BLE) capabilities
- The new 1-Series sensor for enhanced reliability
- Serviceability
- Ease of maintenance
- Lightweight convenience
- Easy-to-read display for multiple languages
Specifications
Class I, Division 1, Groups A, B, C, D T4; Class I, Zone 0, AEx/Ex ia IIC T4 Ga -40°C ≤ Tamb ≤60°C; ATEX: Sira 19ATEX2004; X 0518 g II 1G Ex ia IIC T4 Ga -40°C ≤ Tamb ≤60°C;; g I M1 Ex ia I Ma -40°C ≤ Tamb ≤60°C;; IECEx: IECEx SIR 19.0004; Ex ia IIC T4 Ga / Ex ia I Ma -40°C ≤ Tamb ≤60°C;; FCC ID: SU3BWS1; IC: 20969-BWS1; RED: RE-D Directive 2014/53/EU | |
Detector with specified sensor, stainless-steel alligator clip and concussion-proof housing; Test Cap with 1 ft of hose; Quick Reference Guide | |
3.6 to 4.1 oz. | |
3.6 cm |
7 cm | |
6.7 cm | |
103 to 116 g |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.