Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Bz 423

Click to view available options
Quantity:
10 mg
50 mg
Description
Bz 423 is an ATP synthase inhibitor (IC50 ~5 μM). Binds the oligomycin sensitivity conferring protein (OSCP) component of mitochondrial F1F0-ATPase in vitro resulting in generation of superoxide and apoptosis. Kills primary B and T cell in mice in vivo. Suppresses autoimmunity and prolongs survival in mouse lupus model.
Specifications
Specifications
Chemical Name or Material | 7-Chloro-1,3-dihydro-5-(4-hydroxyphenyl)-1-methyl-3-(2-naphthalenylmethyl)-2H-1,4-benzodiazepin-2-one |
CAS | 216691-95-1 |
Quantity | 10 mg |
Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
Molecular Formula | C27H21ClN2O2 |
Formula Weight | 440.92 |
Physical Form | Solid |
Color | White |
Product Type | Bioactive Small Molecule |
Purity | >98% |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction