Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ C 176
GREENER_CHOICE

Catalog No. p-7199113
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

mSTING antagonist

C 176 is a mSTING antagonist. Covalently binds Cys91 of mouse but not human STING. Blocks activation-induced palmitoylation of STING and formation of STING clusters. Reduces CMA (Cat. No. 6677) -induced increase in serum IFNs in vivo and suppresses systemic inflammation in a mouse inflammatory disease model.

Specifications

CAS 314054-00-7
Quantity 10 mg
Target STING Antagonists
Molecular Formula C11H7IN2O4
Formula Weight 808
Purity 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.